Dimethoxy(dimethyl)silane 95+%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Dimethyldimethoxysilane
351530010
351531000
351530010.EA
7730
CZK
351530010.
351531000.
Dimethoxy(dimethyl)silane 95+%, AcroSeal®
Dimethoxy(dimethyl)silane
Vzorec:
C₄H₁₂O₂Si Bod varu: 81…82 °C (760 mmHg) Bod tání: –80 °C Hustota: 0,865 g/cm³ Bod vzplanutí: –8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025691 CAS číslo: 1112-39-6 EINECS: 214-189-4 UN: 1993 ADR: 3,II |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3690 to 1.3710 (20°C, 589 nm) |
GC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...