Tisk…

3,4-Diethoxy-3-cyclobutene-1,2-dione ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Diethyl squarate, 3,4-Diethoxycyclobut-3-ene-1,2-dione

A19248.03 A19248.06 A19248.14
A19248.03EA 1750 CZK
A19248.03 A19248.06 A19248.14
3,4-Diethoxy-3-cyclobutene-1,2-dione ≥98%
3,4-Diethoxy-3-cyclobutene-1,2-dione

Purity: 98% CAS No.: 5231-87-8 Molecular Formula: C8H10O4 Formula Weight: 170.16 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5090 Boiling Point: 95°/0.1mm Beilstein Reference: 640626 MDL No.: MFCD00001333

Vzorec: C₈H₁₀O₄
MW: 170,16 g/mol
Bod varu: 95 °C (0,1 mmHg)
Hustota: 1,162
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001333
CAS číslo: 5231-87-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR