5-Chloro-2-thiophenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
5-Chlorothiophene-2-sulphonyl chloride
M-Clarity™ quality level = MQ100
544272-1G
544272-5G
SIAL544272-1GEA
1460
CZK
SIAL544272-1G
SIAL544272-5G
5-Chloro-2-thiophenesulphonyl chloride, Sigma-Aldrich®
5-Chloro-2-thiophenesulphonyl chloride
Vzorec:
C₄H₂Cl₂O₂S₂ MW: 217,1 g/mol Bod varu: 64…66 °C (0,05 mmHg) Bod tání: 28 °C Hustota: 1,623 g/cm³ (20 °C) |
MDL Number:
MFCD00051667 CAS číslo: 2766-74-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...