Tisk…

5-Chloro-2-thiophenesulphonyl chloride ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 5-Chlorothiophene-2-sulphonyl chloride

A11348.03 A11348.06 A11348.09
A11348.03EA 519 CZK
A11348.03 A11348.06 A11348.09
5-Chloro-2-thiophenesulphonyl chloride ≥97%
5-Chloro-2-thiophenesulphonyl chloride

Purity: 97% CAS No.: 2766-74-7 Molecular Formula: C4H2Cl2O2S2 Formula Weight: 217.09 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5860 Boiling Point: 117-120°/14mm Beilstein Reference: 130710 UN No.: 3265 MDL No.: MFCD00051667

Vzorec: C₄H₂Cl₂O₂S₂
MW: 217,1 g/mol
Bod varu: 117…120 °C (14 mmHg)
Bod tání: 28 °C
Hustota: 1,623
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00051667
CAS číslo: 2766-74-7
EINECS: 700-248-6
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR