Tisk…

p-Toluamide ≥98%

Dodavatel: Thermo Scientific

Synonyms: 4-Methylbenzamide

A11559.14 A11559.22 A11559.36
A11559.14EA 1210 CZK
A11559.14 A11559.22 A11559.36
p-Toluamide ≥98%
p-Toluamide

Purity: 98+% CAS No.: 619-55-6 Molecular Formula: C8H9NO Formula Weight: 135.17 Melting Point: 158-160° Beilstein Reference: 774353

Vzorec: C₈H₉NO
MW: 135,17 g/mol
Bod varu: 305 °C (1013 hPa)
Bod tání: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00008002
CAS číslo: 619-55-6
EINECS: 210-600-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR