Tisk…

4,5-Dibromo-2-thiophenesulphonyl chloride ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4,5-Dibromothiophene-2-sulphonyl chloride

A13384.06 A13384.03 A13384.14
A13384.06EA 4190 CZK
A13384.06 A13384.03 A13384.14
4,5-Dibromo-2-thiophenesulphonyl chloride ≥97%
4,5-Dibromo-2-thiophenesulphonyl chloride

Purity: 97% CAS No.: 81606-31-7 Molecular Formula: C4HBr2ClO2S2 Formula Weight: 340.45 Melting Point: 75-79° Beilstein Reference: 162960 UN No.: 3261 MDL No.: MFCD00041439

Vzorec: C₄HBr₂ClO₂S₂
MW: 340,44 g/mol
Bod tání: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00041439
CAS číslo: 81606-31-7
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR