Tisk…

m-Toluamide ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Methylbenzamide

B24442.14 B24442.22
B24442.14EA 1390 CZK
B24442.14 B24442.22
m-Toluamide ≥99%
m-Toluamide

Purity: 99% CAS No.: 618-47-3 Molecular Formula: C8H9NO Formula Weight: 135.17 Melting Point: 93-96° Beilstein Reference: 1857374

Vzorec: C₈H₉NO
MW: 135,17 g/mol
Bod tání: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007990
CAS číslo: 618-47-3
EINECS: 210-553-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR