Akrylonitril stabilised, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200
8.00834.0100
8.00834.1000
8.00834.0100EA
1360
CZK
8.00834.0100
8.00834.1000
Akrylonitril stabilised, Sigma-Aldrich®
Akrylonitril
Vzorec:
C₃H₃N Bod varu: 77,3 °C (1013 hPa) Hustota: 0,7951 g/cm³ (20 °C) Bod vzplanutí: –5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001927 CAS číslo: 107-13-1 EINECS: 203-466-5 UN: 1093 ADR: 3 (6.1),I Merck Index: 13,00133 |
Výsledky specifikačního testu
Solubility | 73 g/l (20 °C) |
Melting Point | -83.55 °C |
Molar Mass | 53.06 g/mol |
Boiling Point | 77.3 °C (1013 hPa) |
Vapor Pressure | 124 hPa (20 °C) |
Flash Point | -4.4 °C |
Refractive Index | 1.391 (20 °C, 589 nm) |
Explosion Limit | 2.8 - 28 %(V) |
Density | 0.81 g/cm3 (20 °C) |
pH | 6.0 - 7.5 (50 g/l, H2O, 20 °C) |
Ignition Point | 480 °C |
Synonyms | Acrylic acid nitrile, Vinyl cyanide |
Hill Formula | C3H3N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...