Tisk…

Akrylonitril stabilised, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


Stabilization: 35-40 ppm Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

110213-1L 110213-100ML 110213-18L 110213-2L 110213-4L 110213-500ML 110213-5ML
SIAL110213-1LEA 2450 CZK
SIAL110213-1L SIAL110213-100ML SIAL110213-18L SIAL110213-2L SIAL110213-4L SIAL110213-500ML SIAL110213-5ML
Akrylonitril stabilised, Sigma-Aldrich®
Akrylonitril
Vzorec: C₃H₃N
MW: 53.06
Bod varu: 77.3
Bod tání: –83.55 °C
Hustota: 0.7951 g/cm³ (20 °C)
Bod vzplanutí: –5 °C
MDL Number: MFCD00001927
CAS číslo: 107-13-1
EINECS: 203-466-5
UN: 1093
ADR: 3 I
Merck Index: 13,00133

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR