Akrylonitril ≥99% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 40 ppm
A13058.0F
A13058.AE
A13058.AP
A13058.0FEA
3210
CZK
A13058.0F
A13058.AE
A13058.AP
Akrylonitril ≥99% stabilizovaný
Akrylonitril
Stabilized with ca 40 ppm 4-methoxyphenol
Purity: 99+%
CAS No.: 107-13-1
Molecular Formula: C3H3N
Formula Weight: 53.06
Flash Point: 0°(32°F)
Physical Form: Liquid
Density: 0.806
Refractive Index: 1.3910
Sensitivity: Light Sensitive
Appearance: Colourless
Odour: Pungent
Melting Point: -84°
Boiling Point: 77-78°
Beilstein Reference: 605310
Merck Reference: 14,131
UN No.: 1093
MDL No.: MFCD00001927
Vzorec:
C₃H₃N MW: 53,06 g/mol Bod varu: 77…78 °C Bod tání: –84 °C Hustota: 0,806 Bod vzplanutí: 0 °C (32 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001927 CAS číslo: 107-13-1 EINECS: 203-466-5 UN: 1093 ADR: 3,I Merck Index: 14,131 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...