2-(Dibutylamino)ethanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N,N-Dibutylethanolamine
MQ100
550035-100ML
550035-500ML
SIAL550035-100MLEA
833
CZK
SIAL550035-100ML
SIAL550035-500ML
2-(Dibutylamino)ethanol, Sigma-Aldrich®
2-(Dibutylamino)ethanol
Vzorec:
C₁₀H₂₃NO MW: 173.3 Bod varu: 224 Bod tání: –75 °C Hustota: 0.8624 g/cm³ (20 °C) |
MDL Number:
MFCD00014033 CAS číslo: 102-81-8 EINECS: 203-057-1 UN: 2873 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...