2-(Butylamino)ethanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N-Butylethanolamine
M-Clarity™ quality level = MQ100
471496-2L
471496-500ML
SIAL471496-2LEA
2920
CZK
SIAL471496-2L
SIAL471496-500ML
2-(Butylamino)ethanol, Sigma-Aldrich®
2-(Butylamino)ethanol
Vzorec:
C₆H₁₅NO Bod varu: 198…200 °C (1013 hPa) Hustota: 0,89 g/cm³ (20 °C) |
MDL Number:
MFCD00020570 CAS číslo: 111-75-1 EINECS: 203-904-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...