Tisk…

4-Acetylbenzonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl p-cyanophenyl ketone, 4'-Cyanoacetophenone, 4-Acetylbenzenecarbonitrile

M-Clarity™ quality level = MQ100

154393-25G 154393-5G
SIAL154393-25GEA 6300 CZK
SIAL154393-25G SIAL154393-5G
4-Acetylbenzonitrile, Sigma-Aldrich®
4-Acetylbenzonitrile
Vzorec: C₉H₇NO
MW: 145,16 g/mol
Bod varu: 110…112 °C (1 mmHg)
Bod tání: 57…58 °C
MDL Number: MFCD00001825
CAS číslo: 1443-80-7
EINECS: 215-885-0
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR