Tisk…
Isopropyl-β-D-thiogalaktopyranosid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
I5502-10G I5502-1G I5502-5G
SIALI5502-10GEA 50200 CZK
SIALI5502-10G SIALI5502-1G SIALI5502-5G
Isopropyl-β-D-thiogalaktopyranosid, Sigma-Aldrich®
Isopropyl-β-D-thiogalaktopyranosid
Vzorec: C₉H₁₈O₅S
Bod varu: 402 °C (1013 hPa)
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS číslo: 367-93-1
EINECS: 206-703-0