Tisk…

Lenvatinib mesylate 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 4-(3-Chloro-4-(3-cyclopropylureido)phenoxy)-7-methoxyquinoline-6-carboxamide methanesulphonate

474520010 474522500
474520010.EA 14340 CZK
474520010. 474522500.
Lenvatinib mesylate 98%
Lenvatinib mesylate
Vzorec: C₂₂H₂₃ClN₄O₇S
Storage Temperature: Refrigerator
MDL Number: MFCD18633219
CAS číslo: 857890-39-2

Objednat

Výsledky specifikačního testu

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR