Tisk…
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) 98%
Dodavatel: Thermo Scientific
Synonyms: 3,3,5-Trimethylcyclohexyl 2-hydroxybenzoate, Coppertone, 2-Hydroxybenzoic Acid 3,3,5-Trimethylcyclohexyl Ester, Salicylic Acid 3,3,5-Trimethylcyclohexyl Ester, 3,3,5-Trimethylcyclohexyl 2-Hydroxybenzoate, Homosalate
474510250 474511000
474510250.EA 1120 CZK
474510250. 474511000.
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) 98%
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture)
Vzorec: C₁₆H₂₂O₃
MW: 262,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019377
CAS číslo: 118-56-9
EINECS: 204-260-8
Objednat
Výsledky specifikačního testu
Appearance clear colorless liquid
Infrared spectrum Conforms
HPLC ≥97.5 % sum of isomers