Tisk…

2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid) 90%

Dodavatel: Thermo Fisher Scientific

474500010 474500050
474500010.EA 8130 CZK
474500010. 474500050.
2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid) 90%
2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid)
Vzorec: C₁₈H₂₀N₂O₆
MW: 360.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00004247
CAS číslo: 1170-02-1
EINECS: 214-625-3

Objednat

Výsledky specifikačního testu

Appearance white to light brown crystalline powder or crystals
Infrared spectrum Conforms
Titration Complexometric ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR