Minodronate monohydrate 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Hydroxy-2-(imidazo[1,2-a]pyridin-3-yl)ethylidene-bisphosphonic acid monohydrate, 1-Hydroxy-2-[imidazo[1,2-a]pyridin-3-yl]ethylidene-1,1-diphosphonic Acid, Minodronic Acid, Minodronic acid monohydrate, (1-Hydroxy-2-(imidazo[1,2-a]pyridin-3-yl)ethane-1,1-diyl)diphosphonic acid hydrate
473351000
473355000
473351000.EA
4530
CZK
473351000.
473355000.
Minodronate monohydrate 98%
Minodronate monohydrate
Vzorec:
C₉H₁₂N₂O₇P₂ MW: 322,15 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00890772 CAS číslo: 155648-60-5 UN: 2811 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | white to light pink powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...