Tisk…

Telotristat etiprate 97%

Dodavatel: Thermo Fisher Scientific

473180250 473181000
473180250.EA 3540 CZK
473180250. 473181000.
Telotristat etiprate 97%
Telotristat etiprate
Vzorec: C₃₆H₃₅ClF₃N₇O₆
Storage Temperature: Freezer
MDL Number: MFCD22741520
CAS číslo: 1137608-69-5

Objednat

Výsledky specifikačního testu

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR