Tisk…

Ranolazine dihydrochloride 98%

Dodavatel: Thermo Scientific

Synonyms: N-(2,6-Dimethylphenyl)-2-(4-(2-hydroxy-3-(2-methoxyphenoxy)propyl)piperazin-1-yl)acetamide dihydrochloride

473160010 473160050
473160010.EA 2260 CZK
473160010. 473160050.
Ranolazine dihydrochloride 98%
Ranolazine dihydrochloride
Vzorec: C₂₄H₃₃N₃O₄
MW: 500,46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03788770
CAS číslo: 95635-56-6
Merck Index: 13,08199

Objednat

Výsledky specifikačního testu

Appearance white to almost white powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR