Lycorine HCl 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(1S,2S,3a1S,12bS)-2,3a1,4,5,7,12b-Hexahydro-1H-[1,3]dioxolo[4,5-j]pyrrolo[3,2,1-de]phenanthridine-1,2-diol hydrochloride, Lycorine hydrochloride
472080010
472082500
472080010.EA
10150
CZK
472080010.
472082500.
Lycorine HCl 98%
Lycorine HCl
Vzorec:
C₁₆H₁₇NO₄·HCl MW: 323,8 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00243111 CAS číslo: 2188-68-3 UN: 2811 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | white to light yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 208°C to 212°C |
Loss on drying | ≤5.0 % (105°C, 3 hrs) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...