Olsalazine Sodium 98%
Dodavatel: Thermo Scientific
472060050
472060250
472060050.EA
3240
CZK
472060050.
472060250.
Olsalazine Sodium 98%
Olsalazine Sodium
Vzorec:
C₁₄H₈N₂Na₂O₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00013300 CAS číslo: 6054-98-4 EINECS: 227-975-7 Merck Index: 13,06911 |
Výsledky specifikačního testu
Appearance | yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Loss on drying | ≤2 % (1 g, 150°C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...