Tisk…

Heptaethylene glycol 90+%

Dodavatel: Thermo Fisher Scientific

Synonyms: 3,6,9,12,15,18-Hexaoxaicosane-1,20-diol, PEG-7

472030010 472030050 472030250
472030010.EA 3670 CZK
472030010. 472030050. 472030250.
Heptaethylene glycol 90+%
Heptaethylene glycol
Vzorec: C₁₄H₃₀O₈
MW: 326.39 g/mol
Bod varu: 435°C (760mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00002876
CAS číslo: 5617-32-3
EINECS: 227-040-3

Objednat

Výsledky specifikačního testu

Appearance colorless to light yellow low melting mass
Infrared spectrum Conforms
HPLC ≥90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR