Tisk…

Neotame 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: (S)-3-((3,3-Dimethylbutyl)amino)-4-(((S)-1-methoxy-1-oxo-3-phenylpropan-2-yl)amino)-4-oxobutanoic acid, N-[N-(3,3-Dimethylbutyl)-L-alpha-aspartyl]-L-phenylalanine 1-Methyl ester, N-(N-(3,3-Dimethylbutyl)-L-alpha-aspartyl)-L-phenylalanine 1-methyl ester

471950010 471950050 471950250
471950010.EA 673 CZK
471950010. 471950050. 471950250.
Neotame 98%
Neotame
Vzorec: C₂₀H₃₀N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD09039056
CAS číslo: 165450-17-9

Objednat

Výsledky specifikačního testu

Appearance white to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 80°C to 88°C
Water ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR