Tisk…

Sivelestat sodium 98%

Dodavatel: Thermo Fisher Scientific

471930010 471932500
471930010.EA 9420 CZK
471930010. 471932500.
Sivelestat sodium 98%
Sivelestat sodium
Vzorec: C₂₀H₂₁N₂NaO₇S·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD01937969
CAS číslo: 201677-61-4

Objednat

Výsledky specifikačního testu

Appearance white to almost white solid
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 72°C to 75°C
Water 13.0 to 14.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR