Tisk…

6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone 98%

Dodavatel: Thermo Scientific

Synonyms: 6-(4-Aminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one, Levosimendan dazinone intermediate

471820010 471820050
471820010.EA 776 CZK
471820010. 471820050.
6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone 98%
6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone
Vzorec: C₁₁H₁₃N₃O
MW: 203,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01692712
CAS číslo: 36725-28-7

Objednat

Výsledky specifikačního testu

Appearance light yellow to brown to dark green crystalline powder
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 194°C to 198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR