Nicotinamide riboside chloride 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
3-Carbamoyl-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyridin-1-ium chloride
471650010
471650050
471650010.EA
2630
CZK
471650010.
471650050.
Nicotinamide riboside chloride 98%
Nicotinamide riboside chloride
Vzorec:
C₁₁H₁₅ClN₂O₅ MW: 290.7002 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27952781 CAS číslo: 23111-00-4 |
Výsledky specifikačního testu
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...