20(S)-Protopanaxadiol 96%
Dodavatel: Thermo Fisher Scientific
471190250
471191000
471190250.EA
4270
CZK
471190250.
471191000.
20(S)-Protopanaxadiol 96%
20(S)-Protopanaxadiol
Vzorec:
C₃₀H₅₂O₃ |
MDL Number:
MFCD12032048 CAS číslo: 30636-90-9 |
Výsledky specifikačního testu
Appearance | white to yellow powder or crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥95.0 % |
Water | ≤1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...