Tisk…

Nonaethylene glycol monododecylether (C12E9)

Dodavatel: Thermo Fisher Scientific

Synonyms: Nonaethylene glycol monododecyl ether, Polidocanol, 3,6,9,12,15,18,21,24,27-Nonaoxanonatriacontan-1-ol

471070500 471071000
471070500.EA 4930 CZK
471070500. 471071000.
Nonaethylene glycol monododecylether (C12E9)
Nonaethylene glycol monododecylether (C12E9)
Vzorec: C₃₀H₆₂O₁₀
MW: 582.8073 g/mol
Bod varu: 616°C (760mmHg)
Bod vzplanutí: 244°C
Storage Temperature: Ambient
MDL Number: MFCD00043375
CAS číslo: 3055-99-0
EINECS: 221-284-4

Objednat

Výsledky specifikačního testu

Appearance colorless to white to light yellow waxy low melting solid
Infrared spectrum Conforms
NMR Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR