Tisk…

Perylene Red 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: N,N'-Bis(2,6-diisopropylphenyl)-1,6,7,12-tetraphenoxy-3,4,9,10-perylenetetracarboxylic diimide, 2,9-Bis(2,6-diisopropylphenyl)-5,6,12,13-tetraphenoxyanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone

470970010 470972500
470970010.EA 7390 CZK
470970010. 470972500.
Perylene Red 95%
Perylene Red
Vzorec: C₇₂H₅₈N₂O₈
MW: 1079.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21608638
CAS číslo: 123174-58-3

Objednat

Výsledky specifikačního testu

Appearance red powder
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR