Dimethyl furan-2,5-dicarboxylate 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
470390010
470390050
470390250
470390010.EA
1100
CZK
470390010.
470390050.
470390250.
Dimethyl furan-2,5-dicarboxylate 98%
Dimethyl furan-2,5-dicarboxylate
Vzorec:
C₈H₈O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00092317 CAS číslo: 4282-32-0 |
Výsledky specifikačního testu
Appearance | white to almost white powder or crystals |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Melting point | 108°C to 112°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...