Tisk…

L-O-Phosphoserine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

470240010 470240050 470240250
470240010.EA 1120 CZK
470240010. 470240050. 470240250.
L-O-Phosphoserine 98%
L-O-Phosphoserine
Vzorec: C₃H₈NO₆P
Bod varu: 475,4°C
Storage Temperature: Ambient
MDL Number: MFCD00065935
CAS číslo: 407-41-0
EINECS: 206-986-0
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance white to light yellow crystalline powder
Infrared spectrum Conforms
Titration with NaOH 97.5 to 102.5 %
Melting point 170°C to 174°C
Specific optical rotation +4° to +6° (20°C, 589 nm) (2% in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR