Tisk…

Rasagiline mesylate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (R)-N-Propargyl-1-indanamine Methanesulphonate, (R)-N-(Prop-2-yn-1-yl)-2,3-dihydro-1H-inden-1-amine methanesulphonate, (R)-N-(2-Propyn-1-yl)-1-indanamine Methanesulphonate

470200010 470200050 470202500
470200010.EA 8170 CZK
470200010. 470200050. 470202500.
Rasagiline mesylate 98%
Rasagiline mesylate
Vzorec: C₁₂H₁₃N·CH₄O₃S
MW: 267,34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08460604
CAS číslo: 161735-79-1

Objednat

Výsledky specifikačního testu

Appearance white to yellow to beige crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 155°C to 159°C
Loss on drying ≤11 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR