Varenicline tartrate 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Chantix, 7,8,9,10-Tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline (2R,3R)-2,3-dihydroxysuccinate
470080010
470082500
470080010.EA
8570
CZK
470080010.
470082500.
Varenicline tartrate 98%
Varenicline tartrate
Vzorec:
C₁₃H₁₃N₃·C₄H₆O₆ Bod varu: 400,6°C Storage Temperature: Ambient |
MDL Number:
MFCD08460603 CAS číslo: 375815-87-5 UN: 3077 ADR: 9,III |
Výsledky specifikačního testu
Appearance | white to light orange to beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Water | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...