Tisk…

Dantrolene sodium hemiheptahydrate 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 1-(((5-(4-Nitrophenyl)furan-2-yl)methylene)amino)imidazolidine-2,4-dione, sodium salt hydrate(2:2:7), Sodium Dantrolene, Dantrolene, 1-[(5-(p-Nitrophenyl)-furfurylidene)amino]hydantoin sodium salt, Dantrium

469940050 469940250
469940050.EA 3060 CZK
469940050. 469940250.
Dantrolene sodium hemiheptahydrate 98%
Dantrolene sodium hemiheptahydrate
Vzorec: C₂₈H₃₄N₈Na₂O₁₇
Storage Temperature: Freezer
MDL Number: MFCD00079130
CAS číslo: 24868-20-0

Objednat

Výsledky specifikačního testu

Appearance yellow to orange to brown crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Water 13.0 to 17.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR