Dabigatran Etexilate 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Pradaxa, Hexyl 2-(4-(((5-((3-ethoxy-3-oxopropyl)(pyridin-2-yl)carbamoyl)-1-methyl-1H-benzo[d]imidazol-2-yl)methyl)amino)benzylidene)hydrazinecarboxylate, Ethyl 3-(2-(((4-(N-((hexyloxy)carbonyl)carbamimidoyl)phenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoate, BIBR 953, (E)-ethyl 3-(2-(((4-(N'-((hexyloxy)carbonyl)carbamimidoyl)phenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoate
469930010
469932500
469930010.EA
8870
CZK
469930010.
469932500.
Dabigatran Etexilate 98%
Dabigatran Etexilate
Vzorec:
C₃₄H₄₁N₇O₅ MW: 627.73 g/mol Bod varu: 827°C Storage Temperature: Freezer |
MDL Number:
MFCD22422841 CAS číslo: 211915-06-9 |
Výsledky specifikačního testu
Appearance | white to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 128°C to 133°C |
Water | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...