Tisk…

Piperidin-1-ylacetic acid 95%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Piperidineacetic Acid

469600010 469600050
469600010.EA 3420 CZK
469600010. 469600050.
Piperidin-1-ylacetic acid 95%
Piperidin-1-ylacetic acid
Vzorec: C₇H₁₃NO₂
MW: 143,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00461042
CAS číslo: 3235-67-4

Objednat

Výsledky specifikačního testu

Appearance white to yellow crystalline powder
Infrared spectrum Conforms
Titration with NaOH 94 to 106 %
Melting point 210°C to 216°C
Water ≤2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR