4-Methyl-1-piperazineacetic acid 95%
Dodavatel: Thermo Scientific
|
Varování
|
469570010
469570050
469570010.EA
1830
CZK
469570010.
469570050.
4-Methyl-1-piperazineacetic acid 95%
4-Methyl-1-piperazineacetic acid
Vzorec:
C₇H₁₄N₂O₂ MW: 158,1983 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03444554 CAS číslo: 54699-92-2 |
Výsledky specifikačního testu
Appearance | white to yellow to light brown crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | 94 to 106 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...