Methyl heptadecanoate 99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Heptadecanoic acid methyll ester, Methyl Margarate, Margaric acid methyll ester
469300010
469300050
469300010.EA
1710
CZK
469300010.
469300050.
Methyl heptadecanoate 99%
Methyl heptadecanoate
Vzorec:
C₁₈H₃₆O₂ Bod varu: 152…153°C Bod vzplanutí: >110°C Storage Temperature: Ambient |
MDL Number:
MFCD00009001 CAS číslo: 1731-92-6 EINECS: 217-055-3 |
Výsledky specifikačního testu
Appearance | white to light yellow or colorless powder or liquid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...