Tisk…

Ketorolac tromethamine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Ketorolac tris salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate, Ketorolac tromethamine salt

469170010 469170050
469170010.EA 2630 CZK
469170010. 469170050.
Ketorolac tromethamine 98%
Ketorolac tromethamine
Vzorec: C₁₅H₁₃NO₃·C₄H₁₁NO₃
MW: 376,4037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00887595
CAS číslo: 74103-07-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,05324

Objednat

Výsledky specifikačního testu

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR