Tisk…

Atovaquone 98%

Dodavatel: Thermo Scientific

Synonyms: trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione, 2-(trans-4-(4-Chlorophenyl)cyclohexyl)-3-hydroxynaphthalene-1,4-dione

468760010 468762500
468760010.EA 6510 CZK
468760010. 468762500.
Atovaquone 98%
Atovaquone
Vzorec: C₂₂H₁₉ClO₃
MW: 366,84 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00889188
CAS číslo: 95233-18-4
EINECS: 200-209-9
Merck Index: 13,00471

Objednat

Výsledky specifikačního testu

Appearance yellow to dark yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 221°C to 225°C
Water ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR