Arabinofuranosyl-guanine hydrate
Dodavatel: Thermo Fisher Scientific
|
Varování
|
468631000
468635000
468631000.EA
7210
CZK
468631000.
468635000.
Arabinofuranosyl-guanine hydrate
Arabinofuranosyl-guanine hydrate
Vzorec:
C₁₀H₁₃N₅O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00065486 CAS číslo: 38819-10-2 |
Výsledky specifikačního testu
Appearance | white to almost white crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Water | ≤7 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...