Tisk…

Tranilast

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-[[3-(3,4-Dimethoxyphenyl)-1-oxo-2-propenyl]amino]benzoic Acid, 2-(3-(3,4-Dimethoxyphenyl)acrylamido)benzoic acid, N-(3,4-Dimethoxycinnamoyl)anthranilic Acid

468610010 468610050
468610010.EA 2370 CZK
468610010. 468610050.
Tranilast
Tranilast
Vzorec: C₁₈H₁₇NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00864787
CAS číslo: 53902-12-8
Merck Index: 13,09646

Objednat

Výsledky specifikačního testu

Appearance white to light yellow to green crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Melting point 207°C to 210°C
Titration with TBAH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR