(2-Hydroxybenzyl)diphenylphosphine oxide
Dodavatel: Thermo Fisher Scientific
|
Varování
|
468560010
468560050
468560010.EA
7210
CZK
468560010.
468560050.
(2-Hydroxybenzyl)diphenylphosphine oxide
(2-Hydroxybenzyl)diphenylphosphine oxide
Vzorec:
C₁₉H₁₇O₂P MW: 308.3109 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00406515 CAS číslo: 70127-50-3 |
Výsledky specifikačního testu
Appearance | white to yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Melting point | 175°C to 180°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...