Tisk…

Ferene disodium salt

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Sodium 5,5'-(3-(pyridin-2-yl)-1,2,4-triazine-5,6-diyl)bis(furan-2-sulphonate), Ferene, 3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulphonic acid disodium salt, Disodium 5-[3-pyridin-2-yl-6-(5-sulphonatofuran-2-yl)-1,2,4-triazin-5-yl]furan-2-sulphonate

467750010 467750050 467750250
467750010.EA 2040 CZK
467750010. 467750050. 467750250.
Ferene disodium salt
Ferene disodium salt
Vzorec: C₁₆H₈N₄Na₂O₈S₂
MW: 494.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040462
CAS číslo: 79551-14-7

Objednat

Výsledky specifikačního testu

Appearance white to dark yellow powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Water ≤13 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR