Tisk…

Ezetimibe

Dodavatel: Thermo Scientific

Synonyms: (3R,4S)-1-(4-Fluorophenyl)-3-((S)-3-(4-fluorophenyl)-3-hydroxypropyl)-4-(4-hydroxyphenyl)azetidin-2-one

467690010 467692500
467690010.EA 7800 CZK
467690010. 467692500.
Ezetimibe
Ezetimibe
Vzorec: C₂₄H₂₁F₂NO₃
MW: 409,43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00937872
CAS číslo: 163222-33-1

Objednat

Výsledky specifikačního testu

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Water ≤0.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR