Tisk…

Cilastatin

Dodavatel: Thermo Scientific

Synonyms: (Z)-7-(((R)-2-Amino-2-carboxyethyl)thio)-2-((S)-2,2-dimethylcyclopropane-1-carboxamido)hept-2-enoic acid

467290010 467290050
467290010.EA 5010 CZK
467290010. 467290050.
Cilastatin
Cilastatin
Vzorec: C₁₆H₂₆N₂O₅S
MW: 358,45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00867379
CAS číslo: 82009-34-5
EINECS: 279-875-8
Merck Index: 13,02294

Objednat

Výsledky specifikačního testu

Appearance white to yellow to light beige crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR