Tisk…

Tianeptine sodium salt

Dodavatel: Thermo Scientific

Synonyms: 7-[[3-Chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl]amino]heptanoic acid S,S-dioxide sodium salt

467050010 467052500
467050010.EA 11920 CZK
467050010. 467052500.
Tianeptine sodium salt
Tianeptine sodium salt
Vzorec: C₂₁H₂₄ClN₂NaO₄S
MW: 458,93 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00879851
CAS číslo: 30123-17-2
EINECS: 250-059-3
Merck Index: 13,09496

Objednat

Výsledky specifikačního testu

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Melting point 183°C to 187°C
Water ≤3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR