Ciprofibrate 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-(p-(2,2-Dichlorocyclopropyl)phenoxy)-2-methylpropionic acid, 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropionic Acid, 2-(4-(2,2-Dichlorocyclopropyl)phenoxy)-2-methylpropanoic acid
365030010
365030050
365030010.EA
2890
CZK
365030010.
365030050.
Ciprofibrate 97%
Ciprofibrate
Vzorec:
C₁₃H₁₄Cl₂O₃ MW: 289.15 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00467135 CAS číslo: 52214-84-3 EINECS: 257-744-6 Merck Index: 13,02336 |
Výsledky specifikačního testu
Appearance | white to light beige powder |
Infrared spectrum | Conforms |
HPLC | ≥96 % |
Melting point | 110°C to 118°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...