1,1-Cyclohexanediacetic anhydride 98%
Dodavatel: Thermo Scientific
Synonyms:
3-Oxaspiro[5.5]undecane-2,4-dione, 3,3-Pentamethyleneglutaric Anhydride, 1,1-Cyclohexanediacetic Anhydride, 3-Oxaspiro[5.5]-2,4-undecanedione
358130050
358130250
358130050.EA
1570
CZK
358130050.
358130250.
1,1-Cyclohexanediacetic anhydride 98%
1,1-Cyclohexanediacetic anhydride
Vzorec:
C₁₀H₁₄O₃ MW: 182,22 g/mol Bod varu: 336°C Storage Temperature: Ambient |
MDL Number:
MFCD00040292 CAS číslo: 1010-26-0 |
Výsledky specifikačního testu
Appearance | white to light yellow to light orange crystalline powder or crystals |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Melting point | 67°C to 72°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...